Orders LinkContactUsLinkComponent
Pasteur pipettes, disposable
Pasteur pipettes, disposable
Pasteur pipettes, disposable
Certificates
Pasteur pipettes, disposable
Pasteur pipettes, disposable
4 Options Available Below
Product Details & Documents
These disposable Pasteur pipettes are made of soda glass.
  • Long tapered tip
  • With constriction for cotton plug
  • Available with or without cotton plug
Certifications: ISO 7712
Documents
Product Family Options
Short form
Product Information
  • Capacity
    Ext.Ø
    Length
    Packed
    Availability
    Price
  • HECH40567091
    Pre-plugged
    Pre-plugged
    2 ml
    7 mm
    150 mm
    250/box, 4 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 125,00 €
    125,00 €
    1 * 1.000 ST
       125,00 €
  • HECH40567001
    Non plugged
    Non plugged
    2 ml
    7 mm
    150 mm
    250/box
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 65,80 €
    65,80 €
    1 * 1.000 ST
       65,80 €
Long form
Product Information
  • Capacity
    Ext.Ø
    Length
    Packed
    Availability
    Price
  • HECH40567092
    Pre-plugged
    Pre-plugged
    2 ml
    7 mm
    230 mm
    250/box
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 32,30 €
    32,30 €
    1 * 250 ST
       32,30 €
  • HECH40567002
    Non plugged
    Non plugged
    2 ml
    7 mm
    230 mm
    250/box, 4 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 70,10 €
    70,10 €
    1 * 1.000 ST
       70,10 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.