ContactUsLinkComponent
Flasks acc. to Kjeldahl, with beaded rim
Flasks acc. to Kjeldahl, with beaded rim
Catalog # HECH41870024
undefined
Flasks acc. to Kjeldahl, with beaded rim
Catalog # HECH41870024
Specifications
  • Capacity:
    500 ml
  • Pk:
    1
Specifications
About this item
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.
Certifications: DIN 12360