Orders LinkContactUsLinkComponent
Reading magnifier
Reading magnifier
90×50 mm
Catalog # ESCH2668950
Supplier:   ESCHENBACH OPTIK
Reading magnifier
Catalog # ESCH2668950
Supplier:  ESCHENBACH OPTIK
Reading magnifier
Reading magnifier
90×50 mm
Price
124,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This aplanatic magnifier is suitable for reading and looking at pictures for long periods.
  • Image free from spherical aberration
  • Edge-to-edge and distortion-free image
  • Comfortable thanks to the ergonomic handle
Specifications
  • Magnification:
    2,7×
  • W×D:
    90×50 mm