Soil analysis filter papers, phosphate-free
Phosphate-free smooth filter papers for soil analysis.
MN 280 1/4 folded filters made from acid-washed paper with a high clarification efficiency for determination of micro-nutrients available to vegetation.
MN 619 G for slow filtration, MN 616 G for medium fast filtration and MN 617 G for fast filtration.
- Catalog No:
- MANA440040
- MANA440045
- MANA440050
- MANA540009
- MANA540011
- MANA540005
- MANA540007
- MANA483045
- MANA483050
- MANA440032
- MANA483038
- MANA440038
- MANA483040
- MANA521012
- MANA521018
- MANA521015
- MANA521024
- MANA540012
- MANA483011
- MANA540045
- MANA540050
- MANA494005
- MANA494007
- MANA540032
- MANA540038
- MANA540024
- MANA540027
- MANA540040
- MANA494012
- MANA494024
- MANA483009
- MANA494027
- MANA494015
- MANA494018
- MANA494040
- MANA494045
- MANA494032
- MANA494038
- MANA494009
- MANA494011
- 516-5008
- MANA540015
- 805-4018
- MANA483012
- MANA440011
- MANA440012
- MANA483015
- MANA440007
- MANA440009
- MANA440024
- MANA483027
- MANA483032
- MANA440027
- MANA440015
- MANA483018
- MANA483024
- MANA440018
- MANA440005
- MANA483005
- MANA494050
- MANA483007
- Weight:
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 85 g/m²
- 75 g/m²
- 85 g/m²
- 75 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 85 g/m²
- 75 g/m²
- 75 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Thickness:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,20 mm
- 0,17 mm
- 0,20 mm
- 0,17 mm
- 0,20 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,17 mm
- 0,20 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,20 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,18 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,20 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,20 mm
- 0,17 mm
- 0,17 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- Filtration speed:
- 100 s
- 100 s
- 100 s
- 100 s
- 100 s
- 100 s
- 100 s
- 22 s
- 22 s
- 100 s
- 22 s
- 100 s
- 22 s
- 95 s
- 95 s
- 95 s
- 95 s
- 100 s
- 22 s
- 100 s
- 100 s
- 9 s
- 9 s
- 100 s
- 100 s
- 100 s
- 100 s
- 100 s
- 9 s
- 9 s
- 22 s
- 9 s
- 9 s
- 9 s
- 9 s
- 9 s
- 9 s
- 9 s
- 9 s
- 9 s
- 95 s
- 100 s
- 100 s
- 22 s
- 100 s
- 100 s
- 22 s
- 100 s
- 100 s
- 100 s
- 22 s
- 22 s
- 100 s
- 100 s
- 22 s
- 22 s
- 100 s
- 100 s
- 22 s
- 9 s
- 22 s
Frequently Bought Together