Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PRS
Solid phase extraction cartridges, Bond Elut™ PRS
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PRS
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ PRS
7 Options Available Below
Product Details & Documents
Bond Elut™ PRS is a strong cation exchange sorbent that is also relatively high in polarity. With no appreciable degree of hydrophobicity, in non-polar solvents, PRS is capable of polar and hydrogen bonding interactions. Due to the very low pKa of PRS, a high ionic strength eluent must be used, and so the use of PRS is recommended for weaker cationic species such as pyridinium compounds.
  • Capable of polar and hydrogen bonding interactions
  • No appreciable non-polar interactions
  • Unique selectivity properties
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113012
  • VARI12113038
  • VARI12102074
  • 551-0308
  • VARI12102094
  • VARI12102039
  • VARI12256010
  • Carbon load:
  • 1,7%
  • 1,7%
  • 1,7%
  • 1,7%
  • 1,7%
  • 1,7%
  • 1,7%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113012
    LRC cartridge
    LRC cartridge
    PRS
    60 Å
    40 µm
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 200,00 €
    200,00 €
    1 * 50 ST
       200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113012
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12113038
    LRC cartridge
    LRC cartridge
    PRS
    60 Å
    40 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 247,00 €
    247,00 €
    1 * 50 ST
       247,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113038
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102074
    Straight barrel cartridge
    Straight barrel cartridge
    PRS
    60 Å
    40 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 349,00 €
    349,00 €
    1 * 100 ST
       349,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102074
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g
  • 551-0308
    Straight barrel cartridge
    Straight barrel cartridge
    PRS
    60 Å
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 296,00 €
    296,00 €
    1 * 100 ST
       296,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0308
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102094
    Straight barrel cartridge
    Straight barrel cartridge
    PRS
    60 Å
    40 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 226,00 €
    226,00 €
    1 * 50 ST
       226,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102094
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102039
    Straight barrel cartridge
    Straight barrel cartridge
    PRS
    60 Å
    40 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 261,00 €
    261,00 €
    1 * 50 ST
       261,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102039
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12256010
    Straight barrel cartridge
    Straight barrel cartridge
    PRS
    60 Å
    40 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 221,00 €
    221,00 €
    1 * 30 ST
       221,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256010
    Carbon load
    1,7%
    Endcapped
    No
    Surface area
    500 m²/g