
Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:12092-47-6
:
Formula:C₁₆H₂₄Cl₂Rh₂
MDL Number:MFCD00012415
:493.08 g/mol
EINECS:235-157-6
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together