Order Entry
Belgium
ContactUsLinkComponent
Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98%
Irritant
Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98%
Catalog # STRE45-0380-5G
CAS Number:  
Irritant
Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98%
Catalog # STRE45-0380-5G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :493.08 g/mol
EINECS:235-157-6
 :GHS07
CAS Number:12092-47-6
Formula:C₁₆H₂₄Cl₂Rh₂
MDL Number:MFCD00012415