Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydroxylapatite
Hydroxylapatite, Sigma-Aldrich®
Hydroxylapatite, Sigma-Aldrich®
391947-100
:
Merck Millipore (Calbiochem)
:
Hydroxylapatite, Sigma-Aldrich®
391947-100
:
Merck Millipore (Calbiochem)
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
502.32 g/mol
:
215-145-7
:
1306-06-5
:
12167-74-7
:
Ca₅HO₁₃P₃
:
MFCD00010904