3-Fluoro-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF0272-100MG
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-5.0 to -7.0 deg (C=2, 1mol/L HCl) (calcd.on anh.substance)
- Water:max. 10.0 %
Specification Test Results
About this item
:
:183.1796 g/mol
CAS Number:19883-77-3
Formula:C₉H₁₀FNO₂
MDL Number:MFCD00066450