Tween® 20 (Polysorbate) 2.5% in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:9005-64-5
:
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986
:1227.53 g/mol
Storage Temperature:Ambient
EINECS:500-018-3
Frequently Bought Together