Tetrakis(decyl)ammonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide
CAS Number:14937-42-9
:
Formula:C₄₀H₈₄BrN
MDL Number:MFCD00043166
Molecular Weight:659.0057 g/mol
EINECS:239-013-3
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:86.0 to 90.0 deg-C
Specification Test Results
Frequently Bought Together