Order Entry
Belgium
ContactUsLinkComponent
Tetramethylammonium hydroxide 10% in methanol for photoresist research
Tetramethylammonium hydroxide 10% in methanol for photoresist research
Catalog # TCIAT1201-25ML
Supplier:  TCI
CAS Number:  
undefined
Tetramethylammonium hydroxide 10% in methanol for photoresist research
Catalog # TCIAT1201-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    10.0 to 11.0 w/w%

Specification Test Results

About this item

 :
 :91.15 g/mol
Merck Index:14,09224
Storage Temperature:Ambient
EINECS:200-882-9
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280