Tetramethylammonium hydroxide 10% in methanol
Supplier: TCI
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Merck Index:14,09224
Storage Temperature:Ambient
EINECS:200-882-9
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
Specification Test Results
Frequently Bought Together