Order Entry
Belgium
ContactUsLinkComponent
(S)-(-)-Hydroxypyrrolidine ≥97.0% (by GC, titration analysis)
Irritant
(S)-(-)-Hydroxypyrrolidine ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
(S)-(-)-Hydroxypyrrolidine ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White or Colorless to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together