
Tris(triphenylphosphine)ruthenium(II) chloride ≥90.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15529-49-4
:
Formula:C₅₄H₄₅Cl₂P₃Ru
MDL Number:MFCD00013077
:958.85 g/mol
EINECS:239-569-7
:GHS07
Specification Test Results
- Appearance:Dark green to Dark blue to Black powder to crystal
- Purity (Argentmetric Titration):min. 90.0 %
Specification Test Results
Frequently Bought Together