Order Entry
Belgium
ContactUsLinkComponent
n-Hexane ≥96.0% (by GC) for HPLC
Flammable
Irritant
Health
Environment
n-Hexane ≥96.0% (by GC) for HPLC
Catalog # TCIAH0490-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
n-Hexane ≥96.0% (by GC) for HPLC
Catalog # TCIAH0490-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 96.0 %
  • Evaporation residue:
    max. 0.001 %
  • Absorbance (210nm):
    max. 0.300
  • Absorbance (220nm):
    max. 0.100

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:14,04694
Boiling Pt:68°C
Melting Pt:–95°C
Density:0,659 g/cm³ (20 °C)
Flash Pt:–22°C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520