Order Entry
Belgium
ContactUsLinkComponent
n-Decane ≥99.0% (by GC)
Flammable
Health
n-Decane ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Health
n-Decane ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

Frequently Bought Together