

Sodium dodecyl sulphate (SDS) ≥97.0% (by titrimetric analysis) for ion pair chromatography
: TCI
:151-21-3
:
:CH₃(CH₂)₁₁OSO₃Na
:MFCD00036175
:288.38 g/mol
:14,08636
:207°C
:1,1 g/cm³ (20 °C)
:≥ 100 °C
:Ambient
:205-788-1
:4.1,II
:GHS07
:GHS08
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 97.0 %
- Absorbance (230nm):max. 0.100
- Absorbance (254nm):max. 0.070
- Solubility in Water:almost transparency
Frequently Bought Together