Trifluoroacetic acid ≥99.0% (by titrimetric analysis) sequenation reagent
Catalog # TCIAQ0053-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (Neutralization titration):min. 99.0 %
- Absorbance:max. 0.07 (280nm)
- Aldehyde:max. 0.5 ppm
Specification Test Results
About this item
:
:114.02 g/mol
Merck Index:14,09681
Boiling Pt:71,8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1,5351 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169