Order Entry
Belgium
ContactUsLinkComponent
Butyl acetate ≥99.0% (by GC) for spectrophotometry
Flammable
Irritant
Butyl acetate ≥99.0% (by GC) for spectrophotometry
Catalog # TCIAA0228-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Butyl acetate ≥99.0% (by GC) for spectrophotometry
Catalog # TCIAA0228-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Absorbance (254nm):
    max. 1.000
  • Absorbance (260nm):
    max. 0.300
  • Absorbance (270nm):
    max. 0.080

Specification Test Results

About this item

 :
 :116.16 g/mol
Merck Index:14,01535
Boiling Pt:126°C
Melting Pt:–76 °C
Density:0,875 g/cm³ (20 °C)
Flash Pt:28°C
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
 :GHS02
 :GHS07
CAS Number:123-86-4
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445