


Iodine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:14,05014
Boiling Pt:184,3 °C (1013 hPa)
Melting Pt:114°C
Density:4,9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Gray to Dark blue to Black powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together