Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC) trimethylsilylating agent
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC) trimethylsilylating agent
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC) trimethylsilylating agent
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %
  • High boiling impurity of Silylation agent:
    No impurity higher than Isopropyl myristate (50 ppm) after 5 min + hold up time

Specification Test Results

Related Information

Frequently Bought Together