D-(+)-Maltose monohydrate ≥98.0% (by HPLC)
: TCI
:6363-53-7
:
:C₁₂H₂₂O₁₁·1H₂O
:MFCD00149343
:360.32 g/mol
:14,05714
:160…165 °C
:Ambient
:200-716-5
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+126 to +131 deg (C=10, H2O (add NH3aq (2+3))) (after drying)
- Solubility in Water:almost transparency
- Water:4.8 to 5.8 %
Frequently Bought Together