1-Heptanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier: TCI
Certificates
About this item
CAS Number:22767-50-6
:
Formula:H₃C(CH₂)₅CH₂SO₃Na
MDL Number:MFCD00007543
:202.25 g/mol
Melting Pt:300 °C
Storage Temperature:Ambient
EINECS:245-210-5
Specification Test Results
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Absorbance (210nm):max. 0.400
- Absorbance (230nm):max. 0.050
- Absorbance (254nm):max. 0.040
Specification Test Results
Frequently Bought Together