Order Entry
Belgium
ContactUsLinkComponent
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis)
Irritant
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    171.0 to 175.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together