Order Entry
Belgium
ContactUsLinkComponent
 
Phenylboronic acid (contains varying amounts of Anhydride)
 :  TCI
 
undefined
Phenylboronic acid (contains varying amounts of Anhydride)
 :  TCI

 

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (Neutralization titration):
    97.0 to 117.0 %

 

Frequently Bought Together