Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ LMS
Solid phase extraction cartridges, Bond Elut™ LMS
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ LMS
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ LMS
6 Options Available Below
Product Details & Documents
Bond Elut™ LMS polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that elution of analytes can be achieved with pure organic solvents or HPLC mobile phase-compatible solvent mixtures of low ionic strength. These characteristics are crucial to allow compatibility with LC/MS or other delicate analytical techniques.
  • Ultra clean styrene-divinylbenzene polymer
  • Optimised 75 μm particle size for reproducible flow
  • High capacity and surface area for efficient extraction
Product Family Options
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0337
    Straight barrel cartridge
    Straight barrel cartridge
    LMSA
    300 Å
    75 µm
    1 ml
    25 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 250,00 €
    250,00 €
    1 * 100 ST
       250,00 €
  • 551-0339
    Straight barrel cartridge
    Straight barrel cartridge
    LMSA
    300 Å
    75 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 312,00 €
    312,00 €
    1 * 100 ST
       312,00 €
  • 551-0340
    Straight barrel cartridge
    Straight barrel cartridge
    LMSA
    300 Å
    75 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 232,00 €
    232,00 €
    1 * 50 ST
       232,00 €
  • 551-0341
    Straight barrel cartridge
    Straight barrel cartridge
    LMSA
    300 Å
    75 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 321,00 €
    321,00 €
    1 * 50 ST
       321,00 €
  • 551-0342
    Straight barrel cartridge
    Straight barrel cartridge
    LMSA
    300 Å
    75 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 594,00 €
    594,00 €
    1 * 50 ST
       594,00 €
  • 551-0404
    Straight barrel cartridge
    Straight barrel cartridge
    LMSA
    300 Å
    75 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 351,00 €
    351,00 €
    1 * 30 ST
       351,00 €