Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ ENV
Solid phase extraction cartridges, Bond Elut™ ENV
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ ENV
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ ENV
8 Options Available Below
Product Details & Documents
Bond Elut™ ENV, a PS/DVB polymer, is designed for the extraction of polar organic residues. It contains 125 μm spherical particles with a high degree of cross linking, advantageous for high volume, fast flow-through applications.
  • Provides polar and non-polar modes
  • Strong hydrogen bonding with analytes
  • Resembles unbonded silica in its capabilities
Product Family Options
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0334
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 262,00 €
    262,00 €
    1 * 100 ST
       262,00 €
  • VARI12105013
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 289,00 €
    289,00 €
    1 * 100 ST
       289,00 €
  • 551-0335
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 187,00 €
    187,00 €
    1 * 50 ST
       187,00 €
  • 551-0336
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 225,00 €
    225,00 €
    1 * 50 ST
       225,00 €
  • VARI12255014
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    6 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 181,00 €
    181,00 €
    1 * 30 ST
       181,00 €
  • VARI12105016
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 339,00 €
    339,00 €
    1 * 50 ST
       339,00 €
  • 551-0402
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 207,00 €
    207,00 €
    1 * 30 ST
       207,00 €
  • 551-0403
    Straight barrel cartridge
    Straight barrel cartridge
    ENV
    450 Å
    125 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 284,00 €
    284,00 €
    1 * 30 ST
       284,00 €