ContactUsLinkComponent
Sample dippers
Sample dippers
Catalog # GREI224140_50
Supplier:  Greiner Bio-One
Sample dippers
Catalog # GREI224140_50
Supplier:  Greiner Bio-One
Specifications
  • Capacity:
    100 ml
  • Description:
    Sample dippers
  • Material:
    PS
  • Seq:
    002
  • Environmentally Preferable:
  • Pk:
    50
Specifications
About this item
Sample dippers designed for safe sampling in food manufacturing.
  • Manufactured from chemically inert PS
Suitable for pre-analytical applications.
Packaging: Individually packed.