Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ CN-E
Solid phase extraction cartridges, Bond Elut™ CN-E
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ CN-E
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ CN-E
4 Options Available Below
Product Details & Documents
A medium polarity sorbent with many uses, Bond Elut™ CN-E is ideal for applications in which extremely non-polar compounds would be irreversibly retained on high carbon load sorbents such as C₈ and C₁₈. This endcapped version of the cyano sorbent is best utilised when extracting analytes from an aqueous matrix.
  • Ideal for extracting aqueous analytes
  • Retention in aqueous and organic matrices
  • Useful for many applications
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113033
  • VARI12102064
  • VARI12102007
  • VARI12102034
  • Carbon load:
  • 8,1%
  • 8,1%
  • 8,1%
  • 8,1%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113033
    LRC cartridge
    LRC cartridge
    CN-E
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113033
    Carbon load
    8,1%
    Endcapped
    Yes
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102064
    Straight barrel cartridge
    Straight barrel cartridge
    CN-E
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 293,00 €
    293,00 €
    1 * 100 ST
       293,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102064
    Carbon load
    8,1%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102007
    Straight barrel cartridge
    Straight barrel cartridge
    CN-E
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 318,00 €
    318,00 €
    1 * 100 ST
       318,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102007
    Carbon load
    8,1%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102034
    Straight barrel cartridge
    Straight barrel cartridge
    CN-E
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 244,00 €
    244,00 €
    1 * 50 ST
       244,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102034
    Carbon load
    8,1%
    Endcapped
    Yes
    Surface area
    500 m²/g