Demeter pipettes
Fournisseur: GLASWARENFABRIK KARL HECHT
Jump to:
About this item
Diluting pipettes, soda glass, with ring marks.
- Fused amber graduation 'ASSISTENT-PERMANENT'
- Calibrated for blowing out up to 1 ml
- Other marks are calibrated to delivery ('EX')
Certifications: DIN 12838, DIN 12839
2 Options Available Below
Product Details & Documents
Diluting pipettes, soda glass, with ring marks.
- Fused amber graduation 'ASSISTENT-PERMANENT'
- Calibrated for blowing out up to 1 ml
- Other marks are calibrated to delivery ('EX')
Certifications: DIN 12838, DIN 12839
Product Family Options
Product Information