Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ CH
Solid phase extraction cartridges, Bond Elut™ CH
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ CH
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ CH
5 Options Available Below
Product Details & Documents
Bond Elut™ CH is a mid-polarity sorbent that exhibits unique selectivities for certain analytes. When employed as a non-polar sorbent, CH has the approximate polarity of a C₂ sorbent. Bond Elut™ CH is often a good choice when non-polar sorbents such as C₁₈, C₈ or C₂ do not provide the desired selectivity.
  • Non-polar CH with polarity similar to C₂
  • Retains polar analytes from aqueous matrices
  • Good choice when common non-polar sorbents do not provide the required selectivity
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113032
  • VARI12102006
  • VARI12102033
  • VARI12256005
  • HEWL12256039
  • Carbon load:
  • 9,6%
  • 9,6%
  • 9,6%
  • 9,6%
  • 9,6%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Pore size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113032
    LRC cartridge
    LRC cartridge
    CH
    60 Å
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113032
    Carbon load
    9,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102006
    Straight barrel cartridge
    Straight barrel cartridge
    CH
    60 Å
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102006
    Carbon load
    9,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102033
    Straight barrel cartridge
    Straight barrel cartridge
    CH
    60 Å
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 229,00 €
    229,00 €
    1 * 50 ST
       229,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102033
    Carbon load
    9,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12256005
    Straight barrel cartridge
    Straight barrel cartridge
    CH
    60 Å
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 189,00 €
    189,00 €
    1 * 30 ST
       189,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256005
    Carbon load
    9,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • HEWL12256039
    Straight barrel cartridge
    Straight barrel cartridge
    CH
    60 Å
    12 ml
    2 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 211,00 €
    211,00 €
    1 * 20 ST
       211,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL12256039
    Carbon load
    9,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
This product has been blocked by your organization. Please contact your purchasing department for more information