ContactUsLinkComponent
Cold trap with scealing cap
Cold trap with scealing cap
Catalog # KNFA057955
Supplier:  KNF
Cold trap with scealing cap
Catalog # KNFA057955
Supplier:  KNF
Specifications
  • Pk:
    1
Specifications
About this item
Cold trap made of borosilicate glass 3.3, with GL 45 closing cap.
  • Features hose with threaded caps GL 18