Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ CBA
Solid phase extraction cartridges, Bond Elut™ CBA
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ CBA
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ CBA
8 Options Available Below
Product Details & Documents
CBA is a mid-polarity sorbent and weak cation exchanger (pKa 4,8). It can be used with a wider range of counter-ions than lower pKa sorbents like SCX and will demonstrate easier elution of quaternary amine functionalised analytes.
  • Cation exchange with no need for extreme basic conditions
  • Wider selectivity range provides more eluent options
  • Polar or non-polar depending on matrix or solvent
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113011
  • VARI12113037
  • VARI12102073
  • 551-0307
  • VARI12102097
  • VARI12102124
  • VARI12256009
  • VARI12256058
  • Carbon load:
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Pore size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113011
    LRC cartridge
    LRC cartridge
    CBA
    60 Å
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113011
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12113037
    LRC cartridge
    LRC cartridge
    CBA
    60 Å
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 252,00 €
    252,00 €
    1 * 50 ST
       252,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113037
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102073
    Straight barrel cartridge
    Straight barrel cartridge
    CBA
    60 Å
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 323,00 €
    323,00 €
    1 * 100 ST
       323,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102073
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0307
    Straight barrel cartridge
    Straight barrel cartridge
    CBA
    60 Å
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 301,00 €
    301,00 €
    1 * 100 ST
       301,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0307
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102097
    Straight barrel cartridge
    Straight barrel cartridge
    CBA
    60 Å
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 233,00 €
    233,00 €
    1 * 50 ST
       233,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102097
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102124
    Straight barrel cartridge
    Straight barrel cartridge
    CBA
    60 Å
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 215,00 €
    215,00 €
    1 * 50 ST
       215,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102124
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12256009
    Straight barrel cartridge
    Straight barrel cartridge
    CBA
    60 Å
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 221,00 €
    221,00 €
    1 * 30 ST
       221,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256009
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12256058
    Straight barrel cartridge
    Straight barrel cartridge
    CBA
    60 Å
    12 ml
    2 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 192,00 €
    192,00 €
    1 * 20 ST
       192,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256058
    Carbon load
    7,4%
    Endcapped
    Yes
    Surface area
    500 m²/g