ContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ C18 EWP
 :  VARIAN
Solid phase extraction cartridges, Bond Elut™ C18 EWP
 :  VARIAN
  • Catalog No:
  • VARI12102139
  • VARI12113068
  • VARI12256130
  • VARI12102136
  • VARI12113071
  • Carbon load:
  • 6,0%
  • 6,0%
  • 6,0%
  • 6,0%
  • 6,0%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 80 m²/g
  • 80 m²/g
  • 80 m²/g
  • 80 m²/g
  • 80 m²/g
Frequently Bought Together