Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ C8
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ C8
Supplier:  VARIAN
Certificates
23 Options Available Below
Product Details & Documents
Bond Elut™ C8 is very similar in properties to C18, but is not as retentive for non-polar compounds, due to its shorter hydrocarbon chain, and therefore reduced carbon loading. C8 is an excellent replacement for C18 when analytes are too strongly retained for effective elution. The potential for polar interactions is somewhat higher than for C18 because there is less coverage of the silica surface. These polar interactions are not, however, a significant property of C8.
  • Excellent for strongly-retained analytes
  • Polar interactions not significant
  • Less retentive than C18
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113002
  • VARI12113075
  • VARI12113025
  • VARI12113028
  • 549-0788
  • 549-0807
  • 551-0301
  • 549-0872
  • 549-0798
  • 551-0313
  • 549-0873
  • 551-0316
  • 549-0786
  • 549-0820
  • 549-0825
  • 549-0827
  • 551-0374
  • 551-0381
  • 551-0428
  • VARI75403050
  • 551-0427
  • VARI7540302C
  • VARI7550301C
  • Carbon load:
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • 12,2%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113002
    LRC cartridge
    LRC cartridge
    C8
    60 Å
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 224,00 €
    224,00 €
    1 * 50 ST
       224,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113002
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12113075
    LRC cartridge
    LRC cartridge
    C8
    60 Å
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 148,00 €
    148,00 €
    1 * 50 ST
       148,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113075
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12113025
    LRC cartridge
    LRC cartridge
    C8
    60 Å
    10 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 199,00 €
    199,00 €
    1 * 50 ST
       199,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113025
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12113028
    LRC cartridge
    LRC cartridge
    C8
    60 Å
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 212,00 €
    212,00 €
    1 * 50 ST
       212,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113028
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 549-0788
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 249,00 €
    249,00 €
    1 * 100 ST
       249,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0788
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0807
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    3 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 154,00 €
    154,00 €
    1 * 50 ST
       154,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0807
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0301
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 237,00 €
    237,00 €
    1 * 100 ST
       237,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0301
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0872
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 865,00 €
    865,00 €
    1 * 500 ST
       865,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0872
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0798
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 154,00 €
    154,00 €
    1 * 50 ST
       154,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0798
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0313
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 159,00 €
    159,00 €
    1 * 50 ST
       159,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0313
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0873
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 280,00 €
    1 280,00 €
    1 * 500 ST
       1 280,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0873
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0316
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 186,00 €
    186,00 €
    1 * 50 ST
       186,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0316
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0786
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 120,00 €
    120,00 €
    1 * 30 ST
       120,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0786
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0820
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 173,00 €
    173,00 €
    1 * 30 ST
       173,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0820
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0825
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    20 ml
    5 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 345,00 €
    345,00 €
    1 * 20 ST
       345,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0825
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 549-0827
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    60 Å
    60 ml
    10 g
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 446,00 €
    446,00 €
    1 * 16 ST
       446,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0827
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
Bond Elut™ Jr
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0374
    Bond Elut™ Jr
    Bond Elut™ Jr
    C8
    60 Å
    500 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 354,00 €
    354,00 €
    1 * 100 ST
       354,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0374
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0381
    Bond Elut™ Jr
    Bond Elut™ Jr
    C8
    60 Å
    1 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 381,00 €
    381,00 €
    1 * 100 ST
       381,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0381
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
VersaPlate™ formats
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0428
    Preassembled 96-well plate
    Preassembled 96-well plate
    C8
    60 Å
    40 µm
    25 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 426,00 €
    426,00 €
    1 * 1 ST
       426,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0428
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI75403050
    Preassembled 96-well plate
    Preassembled 96-well plate
    C8
    60 Å
    40 µm
    50 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 448,00 €
    448,00 €
    1 * 1 ST
       448,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI75403050
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0427
    Preassembled 96-well plate
    Preassembled 96-well plate
    C8
    60 Å
    40 µm
    100 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 448,00 €
    448,00 €
    1 * 1 ST
       448,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0427
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI7540302C
    Preassembled 96-well plate
    Preassembled 96-well plate
    C8
    60 Å
    40 µm
    200 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 499,00 €
    499,00 €
    1 * 1 ST
       499,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI7540302C
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI7550301C
    VersaPlate™ tube
    VersaPlate™ tube
    C8
    60 Å
    40 µm
    100 mg
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 314,00 €
    314,00 €
    1 * 96 ST
       314,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI7550301C
    Carbon load
    12,2%
    Endcapped
    Yes
    Surface area
    500 m²/g