Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ C1
Solid phase extraction cartridges, Bond Elut™ C1
 :   VARIAN
Solid phase extraction cartridges, Bond Elut™ C1
 :  VARIAN
Solid phase extraction cartridges, Bond Elut™ C1
Solid phase extraction cartridges, Bond Elut™ C1
Due to the methyl group and subsequent low carbon load, Bond Elut™ C1 is the least retentive of all alkyl group bonded phases for non-polar compounds. However, due to the extensive end capping of this sorbent to mask polar silanol activity, retention and elution of polar and multi-functional analytes can still be achieved. Typical applications include plasma, urine and aqueous samples.
  • Least retentive of all alkyl group bonded phases
  • Easy retention and release of polar compounds
  • Easy retention and release of multi-functional compounds
  • Catalog No:
  • VARI12102004
  • VARI12102090
  • VARI12102031
  • Carbon load:
  • 4,1%
  • 4,1%
  • 4,1%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Product Family Options
  • Description
    Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
  • Straight barrel cartridge
    C1
    60 Å
    40 µm
    1 ml
    100 mg
    1 * 100 ST
    1 * 100 ST - 261,00 €
    261,00 €
    1 * 100 ST
       261,00 €
    Specifications
    Cat. No.
    VARI12102004
    Carbon load
    4,1%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • Straight barrel cartridge
    C1
    60 Å
    40 µm
    3 ml
    100 mg
    1 * 50 ST
    1 * 50 ST - 146,00 €
    146,00 €
    1 * 50 ST
       146,00 €
    Specifications
    Cat. No.
    VARI12102090
    Carbon load
    4,1%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • Straight barrel cartridge
    C1
    60 Å
    40 µm
    3 ml
    500 mg
    1 * 50 ST
    1 * 50 ST - 188,00 €
    188,00 €
    1 * 50 ST
       188,00 €
    Specifications
    Cat. No.
    VARI12102031
    Carbon load
    4,1%
    Endcapped
    Yes
    Surface area
    500 m²/g