Orders LinkContactUsLinkComponent
Micro control valves
Micro control valves
Micro control valves
Certificates
5 Options Available Below
Product Details & Documents
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.
Product Family Options
Product Information
  • Availability
    Price
  • SGEA1236091
    Manual on/off valve, pneumatic (MOVP), "L" single outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
    Manual on/off valve, pneumatic (MOVP), "L" single outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 747,00 €
    747,00 €
    1 * 1 ST
       747,00 €
  • SGEA1236103
    Manual on/off valve, pneumatic (MOVPT), "T" dual outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
    Manual on/off valve, pneumatic (MOVPT), "T" dual outlet, stem length 50 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 797,00 €
    797,00 €
    1 * 1 ST
       797,00 €
  • SGEA1236104
    Manual on/off valve, pneumatic (MOVPTL), "T" dual outlet, stem length 100 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
    Manual on/off valve, pneumatic (MOVPTL), "T" dual outlet, stem length 100 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 797,00 €
    797,00 €
    1 * 1 ST
       797,00 €
  • SGEA1236282
    Brass micro control valve (BMCV-1), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure, flow rate 0 - 2000 ml/min
    Brass micro control valve (BMCV-1), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure, flow rate 0 - 2000 ml/min
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • SGEA1236283
    Stainless steel manual on/off valve (SMOV), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure
    Stainless steel manual on/off valve (SMOV), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Seal seat removal tool
    Seal seat removal tool
    SGEA1236101
    Log in for availability
    1 * 1 ST - 20,40 €
Product Information
  • Catalog #
    Availability
    Price
  • VSV-6 Vespel valve seat
    VSV-6 Vespel valve seat
    SGEA123681
    Log in for availability
    1 * 5 ST - 140,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PTFE seals for MOVP valves
    PTFE seals for MOVP valves
    SGEA123687
    Log in for availability
    1 * 2 ST - 31,90 €