
4-Hydroxy-2-quinolinecarboxylic acid hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:492-27-3
Formula:C₁₀H₇NO₃·xH₂O
MDL Number:MFCD00006753
:189.17 g/mol (anhy)
Merck Index:14,05327
EINECS:207-751-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Water:max. 9.0 %
Specification Test Results
Frequently Bought Together