Order Entry
Belgium
ContactUsLinkComponent
2,2',3,4,4',5,5'-Heptachlorobiphenyl-[13C12] (PCB No. 180) 40 µg/ml in n-nonane (99% 13C), CIL
2,2',3,4,4',5,5'-Heptachlorobiphenyl-[13C12] (PCB No. 180) 40 µg/ml in n-nonane (99% 13C), CIL
Certificates
2,2',3,4,4',5,5'-Heptachlorobiphenyl-[13C12] (PCB No. 180) 40 µg/ml in n-nonane (99% 13C), CIL

Specifications

  • CAS No.:
    35065-29-3
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,2',3,4,4',5,5'-Heptachlorobiphenyl-[13C12] (PCB No. 180), (99% 13C)

Specifications

Frequently Bought Together