Order Entry
Belgium
ContactUsLinkComponent
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥99.0% (by HPLC)
Irritant
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥99.0% (by HPLC)
Catalog # TCIAA0484-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥99.0% (by HPLC)
Catalog # TCIAA0484-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 % (calcd.on anh.substance)
  • Content of Magnesium:
    3.8 to 4.0% (calcd.on anh.substance)
  • Water:
    5.0 to 10.0 %

Specification Test Results

About this item

 :
 :620.98 g/mol (anhy)
EINECS:242-004-7
 :GHS07
CAS Number:18108-68-4
Formula:C₃₂H₂₄MgN₂O₆S₂·xH₂O
MDL Number:MFCD00065353