Order Entry
Belgium
ContactUsLinkComponent
Acetonitrile with 0,1% (v/v) acetic acid
Acetonitrile with 0,1% (v/v) acetic acid
Catalog # ROTHCP01.1
Supplier:  Roth Carl
CAS Number:  
Acetonitrile with 0,1% (v/v) acetic acid
Catalog # ROTHCP01.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :GHS02
 :GHS07
 :75-05-8