p-Tolyl acetate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:140-39-6
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008703
:150.1745 g/mol
Boiling Pt:214°C
Flash Pt:95°C
EINECS:205-413-1
:GHS07
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together