m-Tolyl acetate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:122-46-3
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00041910
:150.18 g/mol
Merck Index:14,02583
Boiling Pt:212°C
Melting Pt:12°C
Flash Pt:95°C
EINECS:204-546-2
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together