
3-Hydroxypyridine-N-oxide ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6602-28-4
:
Formula:C₅H₅NO₂
MDL Number:MFCD00006200
:111.0987 g/mol
EINECS:229-545-4
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 96.0 %
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together