
Indomethacin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53-86-1
:
Formula:C₁₉H₁₆ClNO₄
MDL Number:MFCD00057095
Merck Index:14,04968
ADR:6.1,I
:357.80 g/mol
EINECS:200-186-5
UN:2811
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:158.0 to 162.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together