Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digoxin ≥96.0% (by HPLC)

Digoxin ≥96.0% (by HPLC)

Toxic
Health
Supplier:   TCI
Digoxin ≥96.0% (by HPLC)
Supplier:  TCI
Certificates
Digoxin ≥96.0% (by HPLC)

2 Options Available Below

Product Details & Documents
  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • Merck Index:14,03167
  • Melting Pt:265°C
  • EINECS:244-068-1
  • UN:1544
  • ADR:6.1,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 96.0 area%
  • Specific rotation [a]20/D:
    +10.0 to +13.0 deg (C=10, Pyridine)
  • Solubility in Pyridine:
    almost transparency
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 18,70 €
    18,70 €
    1 * 100 mg
       18,70 €

  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 75,90 €
    75,90 €
    1 * 1 g
       75,90 €