Order Entry
Belgium
ContactUsLinkComponent
 
 
 
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) ≥95.0% (by GC)
 :  TCI
 
undefined
 
 
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together