α-Cyclodextrin ≥98.0% (by HPLC)
Supplier: TCI
About this item
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- Merck Index:14,02718
- EINECS:233-007-4
3 Options Available Below
Product Details & Documents
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- Merck Index:14,02718
- EINECS:233-007-4
Specifications for Product Family
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+146.0 to +153.0 deg (C=1, H2O) (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 10.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



