Order Entry
Belgium
ContactUsLinkComponent
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Orange to Green flake
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Methanolysis Method):
    min. 97.0 %
  • Melting point:
    165.0 to 170.0 deg-C
  • Solubility in Acetone:
    almost transparency

 

Frequently Bought Together